Assay Details
| Gene Symbol: Itga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033228.3 | NP_001028400.2 | 15 - 16 | 2421 | 69 | |
| GenBank mRNA | AK035870.1 | - | 15 - 16 | 2422 | 69 | |
| AK040830.1 | - | 6 - 7 | 845 | 69 | ||
| AK043769.1 | - | 15 - 16 | 2360 | 69 | ||
| AK137169.1 | - | 15 - 16 | 2186 | 69 | ||
| BC014765.1 | - | 15 - 16 | 2285 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itga1
Genomic Map