Assay Details
| Gene Symbol: Itga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033228.3 | NP_001028400.2 | 9 - 10 | 1526 | 67 | |
| GenBank mRNA | AK035870.1 | - | 9 - 10 | 1527 | 67 | |
| AK043769.1 | - | 9 - 10 | 1465 | 67 | ||
| AK137169.1 | - | 9 - 10 | 1291 | 67 | ||
| BC014765.1 | - | 9 - 10 | 1390 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itga1
Genomic Map