Assay Details
| Gene Symbol: Wnt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009520.3 | NP_033546.2 | 3 - 4 | 915 | 69 | |
| GenBank mRNA | AF038384.1 | - | 1 - 2 | 219 | 69 | |
| AF070988.1 | - | 3 - 4 | 792 | 69 | ||
| AK035653.1 | - | 3 - 4 | 916 | 69 | ||
| AK043566.1 | - | 3 - 4 | 1127 | 68 | ||
| BC119276.1 | - | 3 - 4 | 846 | 8734025 | 69 | |
| BC119278.1 | - | 3 - 4 | 846 | 8734027 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wnt2b
Genomic Map