Assay Details
| Gene Symbol: Spn-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033583.1 | 1 - 2 | 569 | 82 | ||
| GenBank mRNA | AK007258.1 | - | 1 - 2 | 570 | 82 | |
| BC048643.1 | - | 1 - 2 | 509 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spn-ps
Genomic Map