Assay Details
| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028149.1 | NP_082425.1 | 12 - 13 | 1190 | 76 | |
| GenBank mRNA | AK012109.1 | - | 12 - 13 | 914 | 76 | |
| AK144786.1 | - | 12 - 13 | 1164 | 76 | ||
| AK220232.1 | - | 12 - 13 | 1143 | 76 | ||
| EF649694.1 | - | 12 - 13 | 937 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl20
Genomic Map