Assay Details
| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028149.1 | NP_082425.1 | 13 - 14 | 1251 | 88 | |
| GenBank mRNA | AK012109.1 | - | 13 - 14 | 975 | 88 | |
| AK144786.1 | - | 13 - 14 | 1225 | 88 | ||
| AK220232.1 | - | 13 - 14 | 1204 | 88 | ||
| EF649694.1 | - | 13 - 14 | 998 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl20
Genomic Map