Assay Details
| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028149.1 | NP_082425.1 | 5 - 6 | 589 | 99 | |
| GenBank mRNA | AK012109.1 | - | 5 - 6 | 313 | 99 | |
| AK144786.1 | - | 5 - 6 | 563 | 99 | ||
| AK161566.1 | - | 5 - 6 | 557 | 99 | ||
| AK220232.1 | - | 5 - 6 | 542 | 99 | ||
| EF649694.1 | - | 5 - 6 | 336 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl20
Genomic Map