Assay Details
| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028149.1 | NP_082425.1 | 6 - 7 | 657 | 118 | |
| GenBank mRNA | AK012109.1 | - | 6 - 7 | 381 | 118 | |
| AK144786.1 | - | 6 - 7 | 631 | 118 | ||
| AK207539.1 | - | 2 - 3 | 161 | 118 | ||
| AK220232.1 | - | 6 - 7 | 610 | 118 | ||
| EF649694.1 | - | 6 - 7 | 404 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl20
Genomic Map