Assay Details
| Gene Symbol: Gpbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122963.1 | NP_001116435.1 | 10 - 11 | 2483 | 114 | |
| NM_028487.4 | NP_082763.3 | 11 - 12 | 2543 | 114 | ||
| GenBank mRNA | AK135728.1 | - | 11 - 12 | 2474 | 114 | |
| AK136894.1 | - | 11 - 12 | 2323 | 114 | ||
| AK145318.1 | - | 10 - 11 | 2472 | 114 | ||
| AK151283.1 | - | 9 - 10 | 113 | |||
| AK163370.1 | - | 11 - 12 | 2391 | 114 | ||
| AK163390.1 | - | 11 - 12 | 2545 | 114 | ||
| AK169406.1 | - | 11 - 12 | 2338 | 114 | ||
| BC016083.1 | - | 9 - 10 | 1337 | 4501399 | 114 | |
| BC067075.1 | - | 10 - 11 | 2257 | 30614988 | 114 | |
| BC076635.1 | - | 10 - 11 | 2313 | 114 | ||
| BC094888.1 | - | 10 - 11 | 2276 | 30628044 | 114 | |
| BC096442.1 | - | 11 - 12 | 2358 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpbp1
Genomic Map