Assay Details
| Gene Symbol: Gpbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122963.1 | NP_001116435.1 | 3 - 4 | 1384 | 114 | |
| NM_028487.4 | NP_082763.3 | 3 - 4 | 1384 | 114 | ||
| GenBank mRNA | AK082704.1 | - | 3 - 4 | 1202 | 114 | |
| AK135728.1 | - | 3 - 4 | 1315 | 114 | ||
| AK145318.1 | - | 3 - 4 | 1373 | 114 | ||
| AK151283.1 | - | 2 - 3 | 605 | 114 | ||
| AK161928.1 | - | 3 - 4 | 1172 | 114 | ||
| AK163370.1 | - | 3 - 4 | 1232 | 114 | ||
| AK163390.1 | - | 3 - 4 | 1386 | 114 | ||
| AK169406.1 | - | 3 - 4 | 1179 | 114 | ||
| AY382529.1 | - | 3 - 4 | 1182 | 114 | ||
| BC016083.1 | - | 2 - 3 | 402 | 4501399 | 114 | |
| BC067075.1 | - | 3 - 4 | 1158 | 30614988 | 114 | |
| BC076635.1 | - | 3 - 4 | 1214 | 114 | ||
| BC094888.1 | - | 3 - 4 | 1177 | 30628044 | 114 | |
| BC096442.1 | - | 3 - 4 | 1201 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC016083.1 transcripts.
Set Membership: |
|

Gpbp1
Genomic Map