Assay Details
| Gene Symbol: Cnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146318.1 | NP_001139790.1 | 1 - 2 | 1009 | 69 | |
| NM_009923.2 | NP_034053.2 | 2 - 3 | 794 | 69 | ||
| GenBank mRNA | AF332055.1 | - | 1 - 2 | 607 | 69 | |
| AF332056.1 | - | 1 - 2 | 607 | 69 | ||
| AK041969.1 | - | 1 - 2 | 1009 | 69 | ||
| AK050628.1 | - | 2 - 3 | 759 | 69 | ||
| AK087415.1 | - | 2 - 3 | 760 | 69 | ||
| AK140280.1 | - | 1 - 2 | 1010 | 69 | ||
| AK154654.1 | - | 2 - 3 | 818 | 69 | ||
| AK158318.1 | - | 2 - 3 | 788 | 69 | ||
| AK158520.1 | - | 2 - 3 | 709 | 69 | ||
| AK172318.1 | - | 2 - 3 | 759 | 69 | ||
| AK172514.1 | - | 2 - 3 | 759 | 69 | ||
| AK212198.1 | - | 1 - 2 | 135 | 69 | ||
| BC005544.1 | - | 2 - 3 | 747 | 3585034 | 69 | |
| BC021904.1 | - | 2 - 3 | 683 | 3669076 | 69 | |
| DQ887820.1 | - | 1 - 2 | 193 | 69 | ||
| M31810.1 | - | 1 - 2 | 1375 | 69 | ||
| M58045.1 | - | 2 - 3 | 717 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnp
Genomic Map