Assay Details
| Gene Symbol: Krt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008470.1 | NP_032496.1 | 4 - 5 | 934 | 80 | |
| GenBank mRNA | AF053235.1 | - | 4 - 5 | 934 | 80 | |
| AK137631.1 | - | 4 - 5 | 995 | 80 | ||
| BC103615.1 | - | 4 - 5 | 934 | 40046209 | 80 | |
| BC103616.1 | - | 4 - 5 | 934 | 40046211 | 80 | |
| BC103617.1 | - | 4 - 5 | 934 | 40046210 | 80 | |
| BC103666.1 | - | 4 - 5 | 934 | 40046212 | 80 | |
| BC114977.1 | - | 4 - 5 | 934 | 40046220 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt16
Genomic Map