Assay Details
| Gene Symbol: Ikzf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011771.1 | NP_035901.1 | 1 - 2 | 293 | 59 | |
| GenBank mRNA | AI549961.1 | - | 1 - 2 | 39 | 59 | |
| AK036657.1 | - | 1 - 2 | 294 | 59 | ||
| AK157177.1 | - | 1 - 2 | 231 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011771.1 transcripts.
Set Membership: |
|

Ikzf3
Genomic Map