Assay Details
| Gene Symbol: Grb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010346.2 | NP_034476.1 | 11 - 12 | 1641 | 69 | |
| GenBank mRNA | AK057640.1 | - | 11 - 12 | 1634 | 69 | |
| AK145996.1 | - | 11 - 12 | 1457 | 69 | ||
| AK168438.1 | - | 11 - 12 | 1425 | 69 | ||
| AK220277.1 | - | 10 - 11 | 1532 | 69 | ||
| BC003295.1 | - | 11 - 12 | 1384 | 3583896 | 69 | |
| BC081547.1 | - | 10 - 11 | 1630 | 69 | ||
| M94450.1 | - | 11 - 12 | 1663 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grb7
Genomic Map