Assay Details
| Gene Symbol: Grb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010346.2 | NP_034476.1 | 13 - 14 | 1790 | 113 | |
| GenBank mRNA | AK057640.1 | - | 13 - 14 | 1783 | 113 | |
| AK145996.1 | - | 13 - 14 | 1606 | 113 | ||
| AK168438.1 | - | 13 - 14 | 1574 | 113 | ||
| AK220277.1 | - | 12 - 13 | 1681 | 113 | ||
| BC003295.1 | - | 13 - 14 | 1533 | 3583896 | 113 | |
| BC081547.1 | - | 12 - 13 | 1779 | 113 | ||
| M94450.1 | - | 13 - 14 | 1812 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grb7
Genomic Map