Assay Details
| Gene Symbol: Cdk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109626.1 | NP_001103096.1 | 8 - 9 | 2817 | 113 | |
| NM_001109628.1 | NP_001103098.1 | 8 - 9 | 2817 | 113 | ||
| NM_026952.2 | NP_081228.2 | 8 - 9 | 2817 | 113 | ||
| GenBank mRNA | AK049376.1 | - | 8 - 9 | 2463 | 113 | |
| AK129237.1 | - | 8 - 9 | 1956 | 113 | ||
| AK137734.1 | - | 6 - 7 | 687 | 113 | ||
| AY072295.1 | - | 8 - 9 | 2808 | 113 | ||
| BC116645.1 | - | 8 - 9 | 2800 | 40086381 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk12
Genomic Map