Assay Details
| Gene Symbol: Psmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009439.1 | NP_033465.1 | 7 - 8 | 1259 | 64 | |
| GenBank mRNA | AK077252.1 | - | 7 - 8 | 1208 | 64 | |
| AK145467.1 | - | 7 - 8 | 1249 | 64 | ||
| AK145488.1 | - | 7 - 8 | 1248 | 64 | ||
| AK153505.1 | - | 7 - 8 | 1176 | 64 | ||
| AK161632.1 | - | 2 - 3 | 143 | 64 | ||
| AK164333.1 | - | 6 - 7 | 1485 | 64 | ||
| AK164593.1 | - | 7 - 8 | 1248 | 64 | ||
| AK165520.1 | - | 7 - 8 | 1194 | 64 | ||
| AK166683.1 | - | 7 - 8 | 1219 | 64 | ||
| AK166714.1 | - | 7 - 8 | 1247 | 64 | ||
| AK167005.1 | - | 7 - 8 | 1234 | 64 | ||
| AK167285.1 | - | 7 - 8 | 1214 | 64 | ||
| AK169690.1 | - | 7 - 8 | 1248 | 64 | ||
| BC003197.1 | - | 7 - 8 | 1259 | 3586427 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmd3
Genomic Map