Assay Details
| Gene Symbol: Stard3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021547.3 | NP_067522.1 | 4 - 5 | 576 | 94 | |
| GenBank mRNA | AK041538.1 | - | 4 - 5 | 532 | 94 | |
| AK150166.1 | - | 4 - 5 | 520 | 94 | ||
| AK150254.1 | - | 4 - 5 | 520 | 94 | ||
| AK155124.1 | - | 4 - 5 | 515 | 94 | ||
| BC003313.1 | - | 4 - 5 | 493 | 3155179 | 94 | |
| X82457.1 | - | 4 - 5 | 560 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard3
Genomic Map