Assay Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021347.4 | NP_067322.1 | 5 - 6 | 745 | 76 | |
| GenBank mRNA | AB033595.1 | - | 5 - 6 | 734 | 76 | |
| AK028645.1 | - | 5 - 6 | 746 | 76 | ||
| AK028698.1 | - | 5 - 6 | 746 | 76 | ||
| AK029062.1 | - | 5 - 6 | 729 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gsdma
Genomic Map