Assay Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003817.1 | NP_001003817.1 | 10 - 11 | 1405 | 68 | |
| GenBank mRNA | AK031099.1 | - | 10 - 11 | 1273 | 68 | |
| AK031542.1 | - | 2 - 3 | 103 | 68 | ||
| AK083669.1 | - | 10 - 11 | 1273 | 68 | ||
| AK129487.1 | - | 10 - 11 | 1375 | 68 | ||
| BC046811.1 | - | 10 - 11 | 1396 | 5702040 | 68 | |
| BC053078.1 | - | 10 - 11 | 1396 | 5702040 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2
Genomic Map