Assay Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003817.1 | NP_001003817.1 | 24 - 25 | 3148 | 111 | |
| GenBank mRNA | AK031099.1 | - | 24 - 25 | 3016 | 111 | |
| AK031542.1 | - | 16 - 17 | 1846 | 111 | ||
| AK083669.1 | - | 23 - 24 | 3098 | 111 | ||
| AK129487.1 | - | 24 - 25 | 3118 | 111 | ||
| AK142296.1 | - | 24 - 25 | 3142 | 111 | ||
| BC023725.1 | - | 1 - 2 | 121 | 111 | ||
| BC027080.1 | - | 6 - 7 | 760 | 111 | ||
| BC046553.1 | - | 10 - 11 | 1503 | 111 | ||
| BC046811.1 | - | 23 - 24 | 3220 | 5702040 | 111 | |
| BC053078.1 | - | 23 - 24 | 3220 | 5702040 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2
Genomic Map