Assay Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003817.1 | NP_001003817.1 | 25 - 26 | 3343 | 69 | |
| GenBank mRNA | AK031099.1 | - | 25 - 26 | 3211 | 69 | |
| AK031542.1 | - | 17 - 18 | 2041 | 69 | ||
| AK083669.1 | - | 24 - 25 | 3293 | 69 | ||
| AK129487.1 | - | 25 - 26 | 3313 | 69 | ||
| AK142296.1 | - | 25 - 26 | 3337 | 69 | ||
| BC023725.1 | - | 2 - 3 | 316 | 69 | ||
| BC027080.1 | - | 7 - 8 | 955 | 69 | ||
| BC046553.1 | - | 11 - 12 | 1698 | 69 | ||
| BC046811.1 | - | 24 - 25 | 3415 | 5702040 | 69 | |
| BC053078.1 | - | 24 - 25 | 3415 | 5702040 | 69 | |
| L47239.1 | - | 1 - 2 | 132 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2
Genomic Map