Assay Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003817.1 | NP_001003817.1 | 3 - 4 | 616 | 89 | |
| GenBank mRNA | AK031099.1 | - | 3 - 4 | 484 | 89 | |
| AK083669.1 | - | 3 - 4 | 484 | 89 | ||
| AK129487.1 | - | 3 - 4 | 586 | 89 | ||
| AK142296.1 | - | 3 - 4 | 609 | 89 | ||
| BC046811.1 | - | 3 - 4 | 607 | 5702040 | 89 | |
| BC053078.1 | - | 3 - 4 | 607 | 5702040 | 89 | |
| CF728071.1 | - | 3 - 4 | 616 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2
Genomic Map