Assay Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003817.1 | NP_001003817.1 | 7 - 8 | 1084 | 75 | |
| GenBank mRNA | AK031099.1 | - | 7 - 8 | 952 | 75 | |
| AK083669.1 | - | 7 - 8 | 952 | 75 | ||
| AK129487.1 | - | 7 - 8 | 1054 | 75 | ||
| AK142296.1 | - | 7 - 8 | 1077 | 75 | ||
| BC046811.1 | - | 7 - 8 | 1075 | 5702040 | 75 | |
| BC053078.1 | - | 7 - 8 | 1075 | 5702040 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2
Genomic Map