Assay Details
Gene Symbol: Arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207231.1 | NP_997114.1 | 3 - 4 | 566 | 70 | |
GenBank mRNA | AK171194.1 | - | 3 - 4 | 577 | 70 | |
BC065791.1 | - | 3 - 4 | 566 | 3824354 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
