Assay Details
| Gene Symbol: Kat2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038010.2 | NP_001033099.1 | 16 - 17 | 2271 | 116 | |
| NM_020004.5 | NP_064388.2 | 16 - 17 | 2274 | 116 | ||
| GenBank mRNA | AF254441.1 | - | 16 - 17 | 2211 | 116 | |
| AK145205.1 | - | 14 - 15 | 2271 | 116 | ||
| AK147042.1 | - | 16 - 17 | 2219 | 116 | ||
| AK150225.1 | - | 16 - 17 | 2252 | 116 | ||
| AK154866.1 | - | 15 - 16 | 2349 | 116 | ||
| AK155347.1 | - | 16 - 17 | 2448 | 116 | ||
| AK155791.1 | - | 16 - 17 | 2273 | 116 | ||
| AK155842.1 | - | 16 - 17 | 2272 | 116 | ||
| AK158079.1 | - | 16 - 17 | 2262 | 116 | ||
| AK179662.1 | - | 4 - 5 | 324 | 116 | ||
| BC003983.1 | - | 1 - 2 | 69 | 116 | ||
| BC011088.1 | - | 16 - 17 | 2181 | 116 | ||
| BC051952.1 | - | 14 - 15 | 2495 | 116 | ||
| BC060050.1 | - | 14 - 15 | 2495 | 116 | ||
| BC063752.1 | - | 16 - 17 | 2256 | 6333090 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kat2a
Genomic Map