Assay Details
| Gene Symbol: Slc1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148938.3 | NP_683740.1 | 5 - 6 | 1138 | 77 | |
| GenBank mRNA | AF330257.1 | - | 4 - 5 | 638 | 77 | |
| AK040798.1 | - | 5 - 6 | 798 | 77 | ||
| AK045949.1 | - | 5 - 6 | 817 | 77 | ||
| AK049090.1 | - | 5 - 6 | 798 | 77 | ||
| AK080879.1 | - | 5 - 6 | 822 | 77 | ||
| AK082736.1 | - | 5 - 6 | 798 | 77 | ||
| AK084968.1 | - | 5 - 6 | 782 | 77 | ||
| AK158777.1 | - | 4 - 5 | 672 | 77 | ||
| BC055300.1 | - | 1 - 2 | 1969 | 77 | ||
| BC058711.1 | - | 5 - 6 | 1061 | 6842500 | 77 | |
| BC066154.1 | - | 5 - 6 | 741 | 30102325 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a3
Genomic Map