Assay Details
| Gene Symbol: Slc1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148938.3 | NP_683740.1 | 7 - 8 | 1656 | 81 | |
| GenBank mRNA | AF330257.1 | - | 6 - 7 | 1156 | 81 | |
| AK031831.1 | - | 7 - 8 | 1319 | 81 | ||
| AK040798.1 | - | 7 - 8 | 1316 | 81 | ||
| AK045949.1 | - | 7 - 8 | 1335 | 81 | ||
| AK049090.1 | - | 7 - 8 | 1316 | 81 | ||
| AK080879.1 | - | 7 - 8 | 1340 | 81 | ||
| AK082736.1 | - | 7 - 8 | 1316 | 81 | ||
| AK084968.1 | - | 7 - 8 | 1300 | 81 | ||
| AK158777.1 | - | 6 - 7 | 1190 | 81 | ||
| BC055300.1 | - | 3 - 4 | 2487 | 81 | ||
| BC058711.1 | - | 7 - 8 | 1579 | 6842500 | 81 | |
| BC066154.1 | - | 7 - 8 | 1259 | 30102325 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a3
Genomic Map