Assay Details
| Gene Symbol: Igf2bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009951.4 | NP_034081.1 | 2 - 3 | 553 | 89 | |
| GenBank mRNA | AF061569.1 | - | 2 - 3 | 372 | 89 | |
| AK044850.1 | - | 2 - 3 | 458 | 89 | ||
| BC051679.1 | - | 2 - 3 | 536 | 30008106 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp1
Genomic Map