| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100012471.mm.1 |
NM_001199293.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012471.mm.1 |
NM_001199293.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012471.mm.1 |
NM_031386.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012471.mm.1 |
NM_031386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012471.mm.1 |
TC1100001336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012472.mm.1 |
NM_001199293.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012472.mm.1 |
NM_001199293.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012472.mm.1 |
NM_031386.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012472.mm.1 |
NM_031386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012472.mm.1 |
TC1100001336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012473.mm.1 |
NM_001199293.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012473.mm.1 |
NM_001199293.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012473.mm.1 |
NM_031386.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012473.mm.1 |
NM_031386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012473.mm.1 |
TC1100001336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012470.mm.1 |
NM_001199293.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012470.mm.1 |
NM_001199293.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012470.mm.1 |
NM_031386.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012470.mm.1 |
NM_031386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012470.mm.1 |
TC1100001336.mm.1 |
MTA TC |