Assay Details
| Gene Symbol: Tex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199293.1 | NP_001186222.1 | 25 - 26 | 4016 | 81 | |
| NM_031386.2 | NP_113563.2 | 25 - 26 | 3997 | 81 | ||
| GenBank mRNA | AK139808.1 | - | 12 - 13 | 1753 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tex14
Genomic Map