Assay Details
| Gene Symbol: Tex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199293.1 | NP_001186222.1 | 11 - 12 | 1462 | 89 | |
| NM_031386.2 | NP_113563.2 | 11 - 12 | 1443 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex14
Genomic Map