Assay Details
| Gene Symbol: Tex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199293.1 | NP_001186222.1 | 12 - 13 | 1644 | 85 | |
| NM_031386.2 | NP_113563.2 | 12 - 13 | 1625 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tex14
Genomic Map