Assay Details
| Gene Symbol: Dhx40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026191.2 | NP_080467.3 | 11 - 12 | 1532 | 154 | |
| GenBank mRNA | AK004610.1 | - | 11 - 12 | 1496 | 154 | |
| AK010512.1 | - | 11 - 12 | 1512 | 154 | ||
| AK029935.1 | - | 11 - 12 | 1503 | 154 | ||
| AK031000.1 | - | 11 - 12 | 1478 | 154 | ||
| AK075991.1 | - | 5 - 6 | 538 | 154 | ||
| AK135592.1 | - | 11 - 12 | 1469 | 154 | ||
| BC058276.1 | - | 11 - 12 | 1450 | 6515479 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx40
Genomic Map