Assay Details
| Gene Symbol: Dhx40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026191.2 | NP_080467.3 | 9 - 10 | 1270 | 59 | |
| GenBank mRNA | AK004610.1 | - | 9 - 10 | 1234 | 59 | |
| AK010512.1 | - | 9 - 10 | 1250 | 59 | ||
| AK029935.1 | - | 9 - 10 | 1241 | 59 | ||
| AK031000.1 | - | 9 - 10 | 1216 | 59 | ||
| AK075991.1 | - | 3 - 4 | 276 | 59 | ||
| AK135592.1 | - | 9 - 10 | 1207 | 59 | ||
| BC058276.1 | - | 9 - 10 | 1188 | 6515479 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx40
Genomic Map