Assay Details
| Gene Symbol: Mtmr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133215.1 | NP_573478.1 | 9 - 10 | 935 | 76 | |
| GenBank mRNA | AF262986.1 | - | 9 - 10 | 935 | 76 | |
| AK146641.1 | - | 9 - 10 | 952 | 76 | ||
| BC058091.1 | - | 9 - 10 | 933 | 6822365 | 76 | |
| BC058364.1 | - | 9 - 10 | 933 | 6820754 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mtmr4
Genomic Map