Assay Details
| Gene Symbol: Papln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205343.1 | NP_001192272.1 | 8 - 9 | 761 | 75 | |
| NM_130887.3 | NP_570957.2 | 8 - 9 | 761 | 75 | ||
| GenBank mRNA | AK053198.1 | - | 8 - 9 | 762 | 75 | |
| AK086767.1 | - | 8 - 9 | 742 | 75 | ||
| AK138980.1 | - | 8 - 9 | 734 | 75 | ||
| BC132475.1 | - | 8 - 9 | 701 | 40130752 | 75 | |
| BC137854.1 | - | 8 - 9 | 701 | 8860873 | 75 | |
| BC144975.1 | - | 8 - 9 | 701 | 9053523 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Papln
Genomic Map