Assay Details
| Gene Symbol: Aldh6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134042.2 | NP_598803.1 | 1 - 2 | 64 | 68 | |
| GenBank mRNA | AF297860.1 | - | 1 - 2 | 64 | 68 | |
| AK033587.1 | - | 1 - 2 | 91 | 68 | ||
| AK082790.1 | - | 1 - 2 | 118 | 68 | ||
| AK128924.1 | - | 1 - 2 | 96 | 68 | ||
| AK147146.1 | - | 1 - 2 | 118 | 68 | ||
| AK155814.1 | - | 1 - 2 | 115 | 68 | ||
| AK164366.1 | - | 1 - 2 | 81 | 68 | ||
| AK169915.1 | - | 1 - 2 | 104 | 68 | ||
| AK170305.1 | - | 1 - 2 | 117 | 68 | ||
| AK171581.1 | - | 1 - 2 | 118 | 68 | ||
| AK171896.1 | - | 1 - 2 | 107 | 68 | ||
| BC033440.1 | - | 87 | 4212084 | 68 | ||
More Information
Set Membership: |
|

Aldh6a1
Genomic Map