Assay Details
| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165256.1 | NP_001158728.1 | 6 - 7 | 783 | 80 | |
| NM_030246.2 | NP_084522.1 | 7 - 8 | 840 | 80 | ||
| GenBank mRNA | AA763635.1 | - | 3 - 4 | 335 | 80 | |
| AK185423.1 | - | 1 - 2 | 49 | 80 | ||
| AK193501.1 | - | 1 - 2 | 49 | 80 | ||
| BC003284.1 | - | 6 - 7 | 749 | 3582095 | 80 | |
| CX208460.1 | - | 1 - 2 | 55 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf4
Genomic Map