Assay Details
| Gene Symbol: Dab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008702.2 | NP_001008702.1 | 8 - 9 | 829 | 75 | |
| NM_001037905.3 | NP_001032994.1 | 8 - 9 | 903 | 75 | ||
| NM_023118.5 | NP_075607.2 | 8 - 9 | 903 | 75 | ||
| GenBank mRNA | AK002850.1 | - | 8 - 9 | 786 | 75 | |
| AK154716.1 | - | 8 - 9 | 887 | 75 | ||
| AK162851.1 | - | 8 - 9 | 874 | 75 | ||
| AK165519.1 | - | 8 - 9 | 787 | 75 | ||
| AK165810.1 | - | 8 - 9 | 784 | 75 | ||
| AK168375.1 | - | 8 - 9 | 877 | 75 | ||
| BC006588.1 | - | 7 - 8 | 724 | 3587226 | 75 | |
| BC016887.1 | - | 8 - 9 | 769 | 4242174 | 75 | |
| CD348957.1 | - | 6 - 7 | 426 | 75 | ||
| U18869.1 | - | 8 - 9 | 836 | 75 | ||
More Information
Set Membership: |
|

Dab2
Genomic Map