Assay Details
| Gene Symbol: Osmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011019.3 | NP_035149.2 | 12 - 13 | 1882 | 105 | |
| GenBank mRNA | AB015978.1 | - | 12 - 13 | 2533 | 105 | |
| AF058805.1 | - | 12 - 13 | 1852 | 105 | ||
| AK157508.1 | - | 7 - 8 | 1055 | 105 | ||
| BC137703.1 | - | 11 - 12 | 1780 | 8860722 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osmr
Genomic Map