Assay Details
| Gene Symbol: Osmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011019.3 | NP_035149.2 | 16 - 17 | 2406 | 72 | |
| GenBank mRNA | AB015978.1 | - | 16 - 17 | 3057 | 72 | |
| AF058805.1 | - | 16 - 17 | 2376 | 72 | ||
| BC137703.1 | - | 15 - 16 | 2304 | 8860722 | 72 | |
More Information
Set Membership: |
|

Osmr
Genomic Map