Assay Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178065.4 | NP_835166.3 | 17 - 18 | 2737 | 70 | |
| GenBank mRNA | AB093230.1 | - | 16 - 17 | 2356 | 70 | |
| AK077015.1 | - | 16 - 17 | 2269 | 70 | ||
| AK133332.1 | - | 16 - 17 | 2273 | 70 | ||
| AK147878.1 | - | 17 - 18 | 2701 | 70 | ||
| AK154529.1 | - | 17 - 18 | 2718 | 70 | ||
| BC006074.1 | - | 8 - 9 | 892 | 70 | ||
| BC049900.1 | - | 11 - 12 | 1427 | 70 | ||
| BC057351.1 | - | 16 - 17 | 2565 | 70 | ||
| BC060658.1 | - | 16 - 17 | 2306 | 30355406 | 70 | |
| BC079590.1 | - | 17 - 18 | 2691 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arel1
Genomic Map