| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200014412.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014412.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014412.mm.1 |
TC1200002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014410.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014410.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014410.mm.1 |
TC1200002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014417.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014417.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014417.mm.1 |
TC1200002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014405.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014405.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014405.mm.1 |
TC1200002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014421.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014421.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014421.mm.1 |
TC1200002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014406.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014406.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014406.mm.1 |
TC1200002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014411.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014411.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014411.mm.1 |
TC1200002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014418.mm.1 |
NM_178065.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014418.mm.1 |
NM_178065.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014418.mm.1 |
TC1200002083.mm.1 |
MTA TC |