Assay Details
| Gene Symbol: Nek9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145138.2 | NP_660120.2 | 5 - 6 | 770 | 92 | |
| GenBank mRNA | AJ489828.1 | - | 5 - 6 | 745 | 92 | |
| AK173323.1 | - | 5 - 6 | 754 | 92 | ||
| BC117971.2 | - | 5 - 6 | 711 | 40098375 | 92 | |
| BC117972.2 | - | 5 - 6 | 710 | 40098378 | 92 | |
More Information
Set Membership: |
|

Nek9
Genomic Map