Assay Details
| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008992.2 | NP_033018.2 | 8 - 9 | 961 | 74 | |
| GenBank mRNA | AJ001166.1 | - | 8 - 9 | 865 | 74 | |
| AK028420.1 | - | 8 - 9 | 999 | 74 | ||
| AK183074.1 | - | 1 - 2 | 65 | 74 | ||
| BC050102.1 | - | 8 - 9 | 1069 | 30012556 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcd4
Genomic Map