Assay Details
| Gene Symbol: Vti1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016800.3 | NP_058080.2 | 1 - 2 | 235 | 117 | |
| GenBank mRNA | AF035821.1 | - | 1 - 2 | 111 | 117 | |
| AK002289.1 | - | 1 - 2 | 227 | 117 | ||
| BC009810.1 | - | 1 - 2 | 165 | 4220215 | 117 | |
| BC080204.1 | - | 1 - 2 | 211 | 3672445 | 117 | |
| CX239415.1 | - | 1 - 2 | 212 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vti1b
Genomic Map