Assay Details
| Gene Symbol: Rdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030017.3 | NP_084293.1 | 2 - 3 | 319 | 59 | |
| GenBank mRNA | AK020927.1 | - | 2 - 3 | 316 | 59 | |
| AK039233.1 | - | 2 - 3 | 320 | 59 | ||
| AK164157.1 | - | 2 - 3 | 321 | 59 | ||
| BC016204.1 | - | 2 - 3 | 302 | 4527851 | 59 | |
| BC067213.1 | - | 2 - 3 | 286 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rdh12
Genomic Map