Assay Details
| Gene Symbol: Rdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030017.3 | NP_084293.1 | 3 - 4 | 470 | 81 | |
| GenBank mRNA | AK020927.1 | - | 3 - 4 | 467 | 81 | |
| AK039233.1 | - | 3 - 4 | 471 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rdh12
Genomic Map