Assay Details
| Gene Symbol: 4921509O07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045501.1 | 2 - 3 | 300 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC067074 | BC067074 | -114.0kb | Anti-sense | Chr. 13 - 113292430 - 113370515 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 34471 |
Gm34471 | 21.9kb | Sense | Chr. 13 - 113507828 - 113568698 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4921509O07Rik
Genomic Map